Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ssu72
Entrez Gene ID
68991
Gene Name
Ssu72 RNA polymerase II CTD phosphatase homolog (yeast)
Gene Aliases
1190002E22Rik, 1500011L16Rik, 2610101M12Rik
Chromosome Location
Chr.4: 155704805 - 155733879 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026899.3 NP_081175.2 1 - 2 647 58
GenBank mRNA AK004446.1 - 1 - 2 337 58
AK005220.1 - 1 - 2 338 58
AK019168.1 - 1 - 2 103 58
AK148131.1 - 1 - 2 411 58
BC016544.1 - 1 - 2 327 58

View Full Product Details