Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mib2
Entrez Gene ID
76580
Gene Name
mindbomb homolog 2 (Drosophila)
Gene Aliases
2210008I11Rik, Zzank1, skd
Chromosome Location
Chr.4: 155654470 - 155669254 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256107.1 NP_001243036.1 13 - 14 2529 72
NM_001256108.2 NP_001243037.2 13 - 14 2349 72
NM_145124.3 NP_660106.2 12 - 13 2433 72
GenBank mRNA AB072336.1 - 12 - 13 1995 72
AK084791.1 - 5 - 6 731 72
AK147940.1 - 13 - 14 2294 72
AK159611.1 - 13 - 14 2246 72
AY974090.1 - 11 - 12 1730 72
BC058086.1 - 10 - 11 2017 72
BC141133.1 - 10 - 11 1822 72
BC145441.1 - 11 - 12 1918 72

View Full Product Details