The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170855.1 | NP_001164326.1 | 9 - 10 | 2082 | 62 |
NM_178872.4 | NP_849203.2 | 9 - 10 | 2083 | 62 | |
GenBank mRNA | AB103063.1 | - | 9 - 10 | 1877 | 62 |
AY251389.1 | - | 9 - 10 | 1840 | 62 | |
BC058106.1 | - | 9 - 10 | 2001 | 62 |