Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spink12
Entrez Gene ID
78242
Gene Name
serine peptidase inhibitor, Kazal type 12
Gene Aliases
9230117E20Rik
Chromosome Location
Chr.18: 44104523 - 44108543 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030061.3 NP_084337.1 1 - 2 250 82
GenBank mRNA AK020352.1 - 1 - 2 151 82
AK136616.1 - 1 - 2 251 82
BC145914.1 - 1 - 2 136 82
BC145916.1 - 1 - 2 136 82
DQ437332.1 - 1 - 2 309 82

View Full Product Details