Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slco1a4
Entrez Gene ID
28250
Gene Name
solute carrier organic anion transporter family, member 1a4
Gene Aliases
AI785519, Oatp1a4, Oatp2, Slc21a5
Chromosome Location
Chr.6: 141805438 - 141856283 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030687.1 NP_109612.1 4 - 5 345 146
GenBank mRNA AB031814.2 - 4 - 5 345 146

View Full Product Details