Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cmas
Entrez Gene ID
12764
Gene Name
cytidine monophospho-N-acetylneuraminic acid synthetase
Gene Aliases
AW208911, CMPNeu5Ac, D6Bwg0250e
Chromosome Location
Chr.6: 142756686 - 142775714 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009908.2 NP_034038.2 3 - 4 640 73
GenBank mRNA AJ006215.1 - 3 - 4 553 73
AK155730.1 - 3 - 4 591 73
BC031500.1 - 3 - 4 569 73
BC063776.1 - 3 - 4 569 73

View Full Product Details