Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
Efcab7
Entrez Gene ID
230500
Gene Name
EF-hand calcium binding domain 7
Gene Aliases
mKIAA1799
Chromosome Location
Chr.4: 99829500 - 99912785 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145549.1 NP_663524.1 - 536 106
GenBank mRNA AK142188.1 - 3 - 3 741 106
AK143777.1 - - 594 106
AK220422.1 - - 485 106
BC020077.1 - - 536 106
BC112327.1 - - 387 106

View Full Product Details