Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Meis3
Entrez Gene ID
17537
Gene Name
Meis homeobox 3
Gene Aliases
AI573393, Mrg2
Chromosome Location
Chr.7: 16174849 - 16186510 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK083907.1 - 9 - 10 1322 67
AK134758.1 - 10 - 11 1093 67
AK155364.1 - 10 - 11 1106 67
BC003762.1 - 10 - 11 1097 67
BC117532.1 - 10 - 11 1069 67
BC118513.1 - 10 - 11 1058 67

View Full Product Details