The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285931.1 | NP_001272860.1 | 11 - 12 | 3279 | 91 |
NM_001285932.1 | NP_001272861.1 | 11 - 12 | 3045 | 91 | |
NM_027883.3 | NP_082159.3 | 11 - 12 | 2770 | 91 | |
GenBank mRNA | AK004733.1 | - | 11 - 12 | 2771 | 91 |
AK007461.1 | - | 2 - 3 | 512 | 91 | |
AK076217.1 | - | 10 - 11 | 2746 | 91 | |
AK129062.1 | - | 11 - 12 | 91 | ||
BC058370.1 | - | 11 - 12 | 3107 | 91 | |
BC080856.1 | - | 11 - 12 | 3045 | 91 |