The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081418.1 | NP_001074887.1 | 13 - 14 | 3731 | 56 |
GenBank mRNA | AK220278.1 | - | 7 - 8 | 4404 | 56 |