The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164224.1 | NP_001157696.1 | 2 - 3 | 153 | 88 |
NM_026877.2 | NP_081153.1 | 3 - 4 | 374 | 88 | |
NM_198223.2 | NP_937866.1 | 2 - 3 | 153 | 88 | |
GenBank mRNA | AK088197.1 | - | 3 - 4 | 225 | 88 |
AK146061.1 | - | 2 - 3 | 154 | 88 | |
AY349132.1 | - | 1 - 2 | 143 | 88 | |
AY349133.1 | - | 3 - 4 | 291 | 88 | |
BC019177.1 | - | 3 - 4 | 317 | 88 | |
BC022115.1 | - | 3 - 4 | 321 | 88 | |
BC031153.1 | - | 3 - 4 | 306 | 88 |