Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cd81
Entrez Gene ID
12520
Gene Name
CD81 antigen
Gene Aliases
Tapa-1, Tapa1, Tspan28
Chromosome Location
Chr.7: 143052750 - 143067930 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133655.2 NP_598416.1 6 - 7 817 73
GenBank mRNA AK136370.1 - 6 - 7 781 73
AK166521.1 - 6 - 7 783 73
AK170741.1 - 6 - 7 782 73
BC011433.1 - 6 - 7 772 73
X59047.1 - 6 - 7 537 73

View Full Product Details