The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252543.1 | NP_001239472.1 | 18 - 19 | 97 | |
NM_001252544.1 | NP_001239473.1 | 18 - 19 | 97 | ||
NM_001252545.1 | NP_001239474.1 | 18 - 19 | 97 | ||
NM_001281816.1 | NP_001268745.1 | 15 - 16 | 97 | ||
NM_019787.4 | NP_062761.2 | 18 - 19 | 97 | ||
GenBank mRNA | AF200326.1 | - | 17 - 18 | 97 | |
AK014597.1 | - | 18 - 19 | 97 | ||
AK029661.1 | - | 15 - 16 | 97 | ||
AK139729.1 | - | 18 - 19 | 97 | ||
AK146083.1 | - | 18 - 19 | 97 | ||
AK149693.1 | - | 18 - 19 | 97 | ||
AK153214.1 | - | 18 - 19 | 97 | ||
AK158398.1 | - | 18 - 19 | 2245 | 97 | |
AK166330.1 | - | 18 - 19 | 2210 | 97 | |
AK167382.1 | - | 18 - 19 | 97 | ||
AK167732.1 | - | 18 - 19 | 97 | ||
BC005464.1 | - | 18 - 19 | 2203 | 97 | |
BC011160.1 | - | 18 - 19 | 2230 | 97 | |
CK619007.1 | - | 2 - 3 | 97 | ||
CK623414.1 | - | 4 - 5 | 97 |