Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Enpp4
- Entrez Gene ID
-
224794
- Gene Name
- ectonucleotide pyrophosphatase/phosphodiesterase 4
- Gene Aliases
-
4933413N07Rik, AA986363, AI195364, E-NPP 4, Gm90, mKIAA0879
- Chromosome Location
-
Chr.17: 44096307 - 44105924 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK157622.1 |
- |
1 - 2 |
56 |
101 |
View Full Product Details