Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc29a1
Entrez Gene ID
63959
Gene Name
solute carrier family 29 (nucleoside transporters), member 1
Gene Aliases
1200014D21Rik, AA407560, ENT1
Chromosome Location
Chr.17: 45585200 - 45599608 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199113.1 NP_001186042.1 8 - 9 931 66
NM_001199114.1 NP_001186043.1 7 - 8 770 66
NM_001199115.1 NP_001186044.1 7 - 8 876 66
NM_001199116.1 NP_001186045.1 7 - 8 836 66
NM_022880.3 NP_075018.1 6 - 7 877 66
GenBank mRNA AA407561.1 - 1 - 2 38 66
AF131212.1 - 6 - 7 878 66
AF305501.1 - 7 - 8 841 66
AK004756.1 - 7 - 8 822 66
AK050089.1 - 7 - 8 861 66
AK146265.1 - 7 - 8 771 66
AK149483.1 - 7 - 8 792 66
AK170463.1 - 7 - 8 808 66
AK181560.1 - 4 - 5 416 66
AK193260.1 - 3 - 4 311 66
AK214211.1 - 3 - 4 311 66
BC004828.1 - 8 - 9 894 66
BC006812.1 - 6 - 7 808 66
EU180577.1 - 6 - 7 746 66

View Full Product Details