The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024856.2 | NP_001020027.1 | 14 - 15 | 2501 | 60 |
NM_001024857.2 | NP_001020028.1 | 13 - 14 | 2427 | 60 | |
NM_080788.3 | NP_542966.2 | 14 - 15 | 2255 | 60 | |
GenBank mRNA | AK046978.1 | - | 14 - 15 | 2474 | 60 |
AK137024.1 | - | 13 - 14 | 2416 | 60 | |
AK148269.1 | - | 13 - 14 | 2431 | 60 | |
AK161858.1 | - | 1 - 2 | 519 | 60 |