The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204973.1 | NP_001191902.1 | 11 - 12 | 3852 | 110 |
NM_010238.3 | NP_034368.2 | 11 - 12 | 3671 | 110 | |
NR_037970.1 | 11 - 12 | 3498 | 110 | ||
GenBank mRNA | AB212273.1 | - | 9 - 10 | 2006 | 110 |
AF045462.1 | - | 10 - 11 | 2817 | 110 | |
AK158970.1 | - | 11 - 12 | 3672 | 110 | |
AK168525.1 | - | 11 - 12 | 3543 | 110 | |
AK220444.1 | - | 10 - 11 | 2819 | 110 | |
BC046809.1 | - | 11 - 12 | 2533 | 110 | |
BC138655.1 | - | 10 - 11 | 2445 | 110 | |
BC138656.1 | - | 10 - 11 | 2445 | 110 | |
KJ650222.1 | - | 10 - 11 | 2137 | 110 |