Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Rnf24
- Entrez Gene ID
-
51902
- Gene Name
- ring finger protein 24
- Gene Aliases
-
2810473M14Rik, 4930505A13Rik, AI317164, C86507, D2Ertd504e
- Chromosome Location
-
Chr.2: 131298488 - 131352892 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK015699.1 |
- |
3 - 4 |
316 |
98 |
View Full Product Details