The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145979.1 | NP_001139451.1 | 11 - 12 | 1713 | 69 |
NM_019581.3 | NP_062527.2 | 11 - 12 | 1748 | 69 | |
GenBank mRNA | AB024573.1 | - | 9 - 10 | 1381 | 69 |
AF168991.1 | - | 11 - 12 | 1653 | 69 | |
AK087511.1 | - | 11 - 12 | 1619 | 69 | |
AK146415.1 | - | 11 - 12 | 1698 | 69 | |
AK148020.1 | - | 11 - 12 | 1670 | 69 | |
BC049089.1 | - | 11 - 12 | 1709 | 69 | |
BC051958.1 | - | 11 - 12 | 1709 | 69 |