Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Klhdc3
Entrez Gene ID
71765
Gene Name
kelch domain containing 3
Gene Aliases
1300011D16Rik, Peas
Chromosome Location
Chr.17: 46674550 - 46680930 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163729.2 NP_001157201.2 3 - 4 610 86
NM_027910.3 NP_082186.2 4 - 5 763 86
GenBank mRNA AB053465.1 - 4 - 5 634 86
AJ278127.1 - 4 - 5 630 86
AK004964.1 - 4 - 5 740 86
AK141673.1 - 3 - 4 595 86
AK170623.1 - 4 - 5 635 86
BC018154.1 - 4 - 5 732 86

View Full Product Details