Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Klhdc3
Entrez Gene ID
71765
Gene Name
kelch domain containing 3
Gene Aliases
1300011D16Rik, Peas
Chromosome Location
Chr.17: 46674550 - 46680930 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163729.2 NP_001157201.2 7 - 8 1104 76
NM_027910.3 NP_082186.2 8 - 9 1257 76
GenBank mRNA AB053465.1 - 8 - 9 1128 76
AJ278127.1 - 8 - 9 1124 76
AK004964.1 - 8 - 9 1234 76
AK141673.1 - 7 - 8 1089 76
AK170623.1 - 8 - 9 1129 76
AK213181.1 - 6 - 7 342 76
BC018154.1 - 8 - 9 1226 76

View Full Product Details