Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Nfya
- Entrez Gene ID
-
18044
- Gene Name
- nuclear transcription factor-Y alpha
- Gene Aliases
-
AA407810, Cbf-b, SEZ-10, SEZ10
- Chromosome Location
-
Chr.17: 48386885 - 48409901 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
M86214.1 |
- |
4 - 5 |
458 |
58 |
View Full Product Details