Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nfya
Entrez Gene ID
18044
Gene Name
nuclear transcription factor-Y alpha
Gene Aliases
AA407810, Cbf-b, SEZ-10, SEZ10
Chromosome Location
Chr.17: 48386885 - 48409901 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110832.1 NP_001104302.1 7 - 8 972 94
NM_010913.2 NP_035043.1 6 - 7 885 94
GenBank mRNA AK004729.1 - 7 - 8 918 94
AK143723.1 - 6 - 7 847 94
AK156487.1 - 7 - 8 802 94
BC029695.1 - 6 - 7 773 94
BC057099.1 - 7 - 8 874 94
M86214.1 - 5 - 6 604 94
M86215.1 - 6 - 7 706 94
X55315.1 - 6 - 7 795 94

View Full Product Details