Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Art1
Entrez Gene ID
11870
Gene Name
ADP-ribosyltransferase 1
Gene Aliases
ADPRT, ARTC1, Yac-1
Chromosome Location
Chr.7: 102101706 - 102111758 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK009284.1 - 2 - 3 154 59

View Full Product Details