Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcne3
Entrez Gene ID
57442
Gene Name
potassium voltage-gated channel, Isk-related subfamily, gene 3
Gene Aliases
2210017H05Rik, MiRP2
Chromosome Location
Chr.7: 100176507 - 100184869 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190870.1 NP_001177799.1 2 - 3 205 91
NM_001190871.2 NP_001177800.1 2 - 3 220 91
GenBank mRNA BC004629.1 - 2 - 3 186 91
DQ832280.1 - 2 - 3 191 91
DV056972.1 - 2 - 3 201 91
EH101376.1 - 2 - 3 233 91

View Full Product Details