Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Plekhb1
Entrez Gene ID
27276
Gene Name
pleckstrin homology domain containing, family B (evectins) member 1
Gene Aliases
AI315628, PHR1, Phret1, evt-1
Chromosome Location
Chr.7: 100643896 - 100662394 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163183.1 NP_001156655.1 1 - 2 166 80
NM_001163184.1 NP_001156656.1 1 - 2 166 80
NM_001291336.1 NP_001278265.1 1 - 2 166 80
GenBank mRNA AK005102.1 - 1 - 2 121 80
AK158172.1 - 1 - 2 157 80
BY284567.1 - 1 - 2 166 80
CX201870.1 - 1 - 2 118 80

View Full Product Details