Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Syt17
Entrez Gene ID
110058
Gene Name
synaptotagmin XVII
Gene Aliases
Bk
Chromosome Location
Chr.7: 118380713 - 118448913 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK135098.1 - 4 - 5 1059 61
AK144560.1 - 6 - 7 1442 61

View Full Product Details