Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Slx1b
- Entrez Gene ID
-
75764
- Gene Name
- SLX1 structure-specific endonuclease subunit homolog B (S. cerevisiae)
- Gene Aliases
-
1110030E23Rik, 2410170E21Rik, 4833422P03Rik, AI853643, Giyd1, Giyd2
- Chromosome Location
-
Chr.7: 126688927 - 126695783 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_033446.1 |
|
1 - 2 |
360 |
81 |
GenBank mRNA |
AK053901.1 |
- |
1 - 2 |
301 |
81 |
BC145468.1 |
- |
1 - 2 |
224 |
81 |
View Full Product Details