The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142744.1 | NP_001136216.1 | 8 - 9 | 685 | 103 |
NM_001142745.1 | NP_001136217.1 | 8 - 9 | 685 | 103 | |
GenBank mRNA | AK011750.2 | - | 8 - 9 | 661 | 103 |
AK011838.1 | - | 8 - 9 | 674 | 103 | |
AK014265.1 | - | 8 - 9 | 659 | 103 | |
AK028297.1 | - | 8 - 9 | 675 | 103 | |
AK133694.1 | - | 8 - 9 | 662 | 103 | |
BC028847.1 | - | 8 - 9 | 664 | 103 |