The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011993.4 | NP_036123.3 | 1 - 2 | 162 | 69 |
GenBank mRNA | AB006715.1 | - | 1 - 2 | 91 | 69 |
AK090197.1 | - | 1 - 2 | 136 | 69 | |
AK165694.1 | - | 1 - 2 | 163 | 69 | |
BC046333.1 | - | 1 - 2 | 136 | 69 | |
BC090674.1 | - | - | 135 | 69 | |
BC138574.1 | - | 1 - 2 | 80 | 69 | |
Y09079.1 | - | 1 - 2 | 141 | 69 |