Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pddc1
Entrez Gene ID
213350
Gene Name
Parkinson disease 7 domain containing 1
Gene Aliases
BC023835, D230016J19Rik
Chromosome Location
Chr.7: 141408184 - 141414125 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172116.4 NP_742114.3 6 - 7 607 109
GenBank mRNA AK081187.1 - 6 - 7 567 109
AK084273.1 - 6 - 7 563 109
AK088159.1 - 6 - 7 583 109
AK140975.1 - 6 - 7 558 109
AK150323.1 - 4 - 5 1277 109
AK150354.1 - 4 - 5 1277 109
AK151259.1 - 4 - 5 1277 109
AK151794.1 - 4 - 5 1277 109
AK151959.1 - 4 - 5 1277 109
AK153110.1 - 4 - 5 1277 109
AK161205.1 - 5 - 6 1410 109
BC023835.1 - 6 - 7 577 109

View Full Product Details