Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tap2
Entrez Gene ID
21355
Gene Name
transporter 2, ATP-binding cassette, sub-family B (MDR/TAP)
Gene Aliases
ABC18, AI462429, APT2, Abcb3, Ham-2, Ham2, MTP2, PSF2, RING11, Tap-2, Y1, jas
Chromosome Location
Chr.17: 34204085 - 34216321 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011530.3 NP_035660.3 7 - 8 1426 61
GenBank mRNA AK020022.1 - 6 - 7 1502 61
AK079667.1 - 7 - 8 1340 61
AK145234.1 - 7 - 8 1386 61
AK151376.1 - 7 - 8 1336 61
AK151483.1 - 7 - 8 1336 61
AK153287.1 - 7 - 8 1336 61
AK159200.1 - 7 - 8 1392 61
BC005578.1 - 7 - 8 1326 61
BC051257.1 - 7 - 8 1339 61
BC057668.1 - 7 - 8 1713 61
CT010387.1 - 6 - 7 1273 61
M90459.1 - 6 - 7 1273 61
U60087.1 - 6 - 7 1276 61
U60089.1 - 6 - 7 1276 61
U60090.1 - 6 - 7 1276 61
U60091.1 - 6 - 7 1276 61
U60092.1 - 6 - 7 1273 61

View Full Product Details