Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trim26
Entrez Gene ID
22670
Gene Name
tripartite motif-containing 26
Gene Aliases
AI462198, Zfp173, Zfp1736
Chromosome Location
Chr.17: 36837124 - 36859398 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025599.3 NP_001020770.2 5 - 6 1020 108
NM_001286726.1 NP_001273655.1 5 - 6 1039 108
NM_001286727.1 NP_001273656.1 5 - 6 988 108
GenBank mRNA AK042252.1 - 5 - 6 1003 108
AK148247.1 - 5 - 6 926 108
BC037110.1 - 5 - 6 1002 108

View Full Product Details