The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025599.3 | NP_001020770.2 | 9 - 10 | 1222 | 68 |
NM_001286726.1 | NP_001273655.1 | 9 - 10 | 1241 | 68 | |
NM_001286727.1 | NP_001273656.1 | 9 - 10 | 1190 | 68 | |
GenBank mRNA | AK042252.1 | - | 9 - 10 | 1205 | 68 |
AK148247.1 | - | 9 - 10 | 1128 | 68 | |
BC026517.1 | - | 4 - 5 | 266 | 68 | |
BC037110.1 | - | 9 - 10 | 1204 | 68 |