Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ipo7
Entrez Gene ID
233726
Gene Name
importin 7
Gene Aliases
A330055O14Rik, C330016G14, Imp7, Ranbp7
Chromosome Location
Chr.7: 110018227 - 110056610 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181517.3 NP_852658.2 20 - 21 2265 67
GenBank mRNA AK039534.1 - 20 - 21 2417 67
AY316153.1 - 20 - 21 2265 67
BC053524.1 - 17 - 18 1948 67
BC064825.1 - 11 - 12 1221 67

View Full Product Details