Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fam98b
Entrez Gene ID
68215
Gene Name
family with sequence similarity 98, member B
Gene Aliases
2610510H03Rik
Chromosome Location
Chr.2: 117249711 - 117271540 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026620.3 NP_080896.2 5 - 6 642 93
GenBank mRNA BC048087.1 - 5 - 6 632 93

View Full Product Details