The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175132.3 | NP_780341.1 | 1 - 2 | 203 | 153 |
GenBank mRNA | AK053054.1 | - | 1 - 2 | 203 | 153 |