Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tbc1d10b
Entrez Gene ID
68449
Gene Name
TBC1 domain family, member 10b
Gene Aliases
1110003P22Rik, C87963
Chromosome Location
Chr.7: 127197459 - 127208468 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144522.5 NP_653105.3 2 - 3 1118 76
GenBank mRNA AF285112.1 - 2 - 3 476 76
AK047320.1 - 2 - 3 1119 76
AK047327.1 - 2 - 3 1119 76
BC072576.1 - 2 - 3 661 76

View Full Product Details