The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019657.4 | NP_062631.1 | 10 - 11 | 877 | 72 |
GenBank mRNA | AF064635.1 | - | 10 - 11 | 856 | 72 |
AK012103.1 | - | 3 - 4 | 291 | 72 | |
AK078841.1 | - | 10 - 11 | 878 | 72 | |
AK081869.1 | - | 10 - 11 | 871 | 72 | |
AK082715.1 | - | 10 - 11 | 871 | 72 | |
AK088521.1 | - | 10 - 11 | 871 | 72 | |
AK150849.1 | - | 10 - 11 | 872 | 72 | |
AK152489.1 | - | 10 - 11 | 872 | 72 | |
AK166027.1 | - | 10 - 11 | 871 | 72 | |
AK167908.1 | - | 10 - 11 | 871 | 72 | |
AK188070.1 | - | 5 - 6 | 354 | 72 | |
AK212469.1 | - | 3 - 4 | 267 | 72 | |
BC010749.1 | - | 10 - 11 | 852 | 72 | |
BC037620.1 | - | 9 - 10 | 794 | 72 | |
BC090659.1 | - | 10 - 11 | 847 | 72 |