The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175467.3 | NP_780676.1 | 5 - 6 | 814 | 62 |
GenBank mRNA | AK048374.1 | - | 4 - 5 | 675 | 62 |
AK054240.1 | - | 4 - 5 | 901 | 62 | |
AK078679.1 | - | 4 - 5 | 878 | 62 | |
AK078686.1 | - | 5 - 6 | 815 | 62 | |
BC094496.1 | - | 4 - 5 | 660 | 62 |