Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc5a7
Entrez Gene ID
63993
Gene Name
solute carrier family 5 (choline transporter), member 7
Gene Aliases
CHT1
Chromosome Location
Chr.17: 54273590 - 54299041 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022025.4 NP_071308.2 1 - 2 453 68
GenBank mRNA AK034415.1 - 1 - 2 337 68
AK053063.1 - 1 - 2 455 68

View Full Product Details