Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trem1
Entrez Gene ID
58217
Gene Name
triggering receptor expressed on myeloid cells 1
Gene Aliases
-
Chromosome Location
Chr.17: 48232367 - 48246924 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021406.5 NP_067381.1 1 - 2 110 80
GenBank mRNA AF241219.1 - 1 - 2 65 80
AK089439.1 - 1 - 2 79 80
AK171753.1 - 1 - 2 79 80
BY203861.1 - 1 - 2 92 80

View Full Product Details