Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mki67
Entrez Gene ID
17345
Gene Name
antigen identified by monoclonal antibody Ki 67
Gene Aliases
D630048A14Rik, Ki-67, Ki67
Chromosome Location
Chr.7: 135689782 - 135716408 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081117.2 NP_001074586.2 13 - 14 9727 70
GenBank mRNA AK133964.1 - 2 - 3 2251 70
BC053453.1 - 2 - 3 2353 70
X82786.1 - 14 - 15 9115 70

View Full Product Details