The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011538.2 | NP_035668.2 | 7 - 8 | 1123 | 68 |
GenBank mRNA | BC140951.1 | - | 6 - 7 | 1126 | 68 |