The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028370.1 | NP_082646.1 | 2 - 3 | 157 | 123 |
GenBank mRNA | AK013364.1 | - | 2 - 3 | 158 | 123 |
AK045286.1 | - | 2 - 3 | 108 | 123 | |
AK050727.1 | - | 2 - 3 | 156 | 123 | |
AK165040.1 | - | 2 - 3 | 134 | 123 | |
BB642981.1 | - | 2 - 3 | 108 | 123 |