The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199733.1 | NP_001186662.1 | 5 - 6 | 1753 | 86 |
NM_007829.4 | NP_031855.3 | 5 - 6 | 1653 | 86 | |
GenBank mRNA | AF006040.1 | - | 4 - 5 | 1504 | 86 |
AK133863.1 | - | 5 - 6 | 1561 | 86 | |
AK144332.1 | - | 5 - 6 | 1586 | 86 | |
AK145906.1 | - | 5 - 6 | 1584 | 86 | |
AK151279.1 | - | 5 - 6 | 1574 | 86 | |
AK153149.1 | - | 5 - 6 | 1541 | 86 | |
AK156425.1 | - | 5 - 6 | 1751 | 86 | |
AK172616.1 | - | 5 - 6 | 1754 | 86 | |
AK213512.1 | - | 1 - 2 | 80 | 86 | |
AY734459.1 | - | 3 - 4 | 1217 | 86 | |
AY734463.1 | - | 2 - 3 | 231 | 86 | |
AY734465.1 | - | 3 - 4 | 1128 | 86 | |
AY734466.1 | - | - | 1408 | 86 | |
BC128373.1 | - | 5 - 6 | 1563 | 86 |