Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tapbp
Entrez Gene ID
21356
Gene Name
TAP binding protein
Gene Aliases
D17Wsu91e, TPN
Chromosome Location
Chr.17: 33919478 - 33929290 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025313.1 NP_001020484.1 2 - 3 429 113
NM_009318.2 NP_033344.1 2 - 3 429 113
GenBank mRNA AF043943.1 - 2 - 3 287 113
AF106278.1 - 2 - 3 219 113
AJ316613.2 - 2 - 3 219 113
AK014566.1 - 2 - 3 284 113
AK077299.1 - 2 - 3 253 113
AK151134.1 - 2 - 3 250 113
AK151807.1 - 2 - 3 232 113
AK151878.1 - 2 - 3 246 113
AK152019.1 - 2 - 3 245 113
AK169697.1 - 2 - 3 250 113
AK170528.1 - 2 - 3 240 113
AY734468.1 - 2 - 3 203 113
AY734469.1 - 2 - 3 202 113
AY734471.1 - 2 - 3 217 113
AY734475.1 - 1 - 2 185 113
AY734477.1 - 2 - 3 204 113
BC015074.1 - 2 - 3 242 113
CF168841.1 - 2 - 3 429 113
KJ650208.1 - 2 - 3 219 113

View Full Product Details