Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vps52
Entrez Gene ID
224705
Gene Name
vacuolar protein sorting 52 (yeast)
Gene Aliases
ARE1, D130068D18, D430041K17Rik, SAC2, Sacm2l, tclw5
Chromosome Location
Chr.17: 33955785 - 33966495 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC063329.1 - 7 - 8 769 74
KJ650214.1 - 8 - 9 795 74

View Full Product Details