Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Hs3st2
- Entrez Gene ID
-
195646
- Gene Name
- heparan sulfate (glucosamine) 3-O-sulfotransferase 2
- Gene Aliases
-
6430516N12Rik, A830061E14Rik, AW491345
- Chromosome Location
-
Chr.7: 121391610 - 121501768 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK043969.1 |
- |
1 - 2 |
58 |
144 |
View Full Product Details