Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dlk2
Entrez Gene ID
106565
Gene Name
delta-like 2 homolog (Drosophila)
Gene Aliases
AI413481, DLK-2, Egfl9
Chromosome Location
Chr.17: 46297248 - 46303271 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286013.1 NP_001272942.1 1 - 2 149 90
GenBank mRNA BG868693.1 - 1 - 2 132 90
FM180474.1 - 1 - 2 149 90

View Full Product Details